Order Entry
Germany
ContactUsLinkComponent
 
Kieselguhr, AnalaR NORMAPUR®, purified and calcined
Catalog # 89056.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Kieselguhr, AnalaR NORMAPUR®, purified and calcined
Catalog # 89056.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility in water:
    max. 0.2%
  • Soluble in acid:
    max. 2.0%
  • Arsenic eliminated by washing:
    max. 0.001%
  • Lead eliminated by washing:
    max. 0.001%
  • Loss on drying (105 °C):
    max. 0.5%
  • Residue on ignition (980 °C):
    max. 2.0%
  • Limit of nonsiliceous substances (max 50 mg):
    passes test

Specification Test Results

About this item

 :
 :60.08 g/mol
Boiling Pt:2230 °C
Melting Pt:1713 °C
Density:2.21 g/cm3
Storage Temperature:Ambient
EINECS:272-489-0
CAS Number:68855-54-9
 :7631-86-9
Formula:O₂Si
MDL Number:MFCD00132803