ContactUsLinkComponent
7000 Safety spectacle, Junior Stealth™
7000 Safety spectacle, Junior Stealth™
Catalog # JSPAASA918-321-100
Supplier:  JSP
undefined
7000 Safety spectacle, Junior Stealth™
Catalog # JSPAASA918-321-100
Supplier:  JSP
Specifications
  • Description:
    Junior Stealth™ 7000 Safety spectacle
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Contour design and flexibility of frame give a good versatile fit onto both smaller and larger faces.
  • One-piece lens
  • Permanently attached side arms for durability
  • Optical class 1
The one-piece lens features permanently attached side arms for durability. Lens and frame offer protection against low energy, high-speed particles at 45 m/s (101 mph). The optical qualities of the lens comply with the requirements of optical class 1 (highest).
Certifications: Conforms to EN 166.