M9400 Wraplite Safety spectacle
Supplier: JSP
Certificates
About this item
Single lens, wraparound, comfortable and lightweight lens design provides excellent field of vision. A popular choice of spectacle that can be worn for long periods of time with continual user comfort ensured.
- Lightweight
- Wraparound design
- Blocks 99,9% of ultraviolet radiation
- Anti-scratch and anti-mist models available
Single lens wraparound safety spectacles offer protection against low energy, high-speed particles at 45 m/s (101 mph). Anti-scratch and anti-mist treatments are available to provide increased lens efficiency.
Certifications: Conforms to EN 166.1.F.