ContactUsLinkComponent
Safety spectacle, Galactus™
Safety spectacle, Galactus™
  JSPAASA480-0A7-600
 :  JSP
undefined
Safety spectacle, Galactus™
  JSPAASA480-0A7-600
 :  JSP
  • Description:
    Galactus™ Safety spectacle
  • Seq:
    01
  • Lenses:
    Clear PC
  • Pk:
    1
Adaptable and ultra-light safety spectacle with base ten lens.
  • Stylish premium safety eyewear
  • Ultra-light TPR frame
  • Adaptable nose bridge
  • Base ten lens
  • Soft touch arms
Galactus™ premium safety eyewear features a base 10 curved lens which follows facial contours perfectly, a unique mouldable nose bridge system that can be adjusted to accommodate all high/low/thin or wide nose bridge profiles, and a frame that utilises a fusion of polymer and TPE to be both lightweight and super strong.
Soft touch arms with elastomer-injected sleeve for more comfort and better adhesion. Polymer inner structure for added flexibility and comfort. The base ten lens provides maximum field of view and more protective coverage.
 : EN 166.