ContactUsLinkComponent
Belt bag, Force®8
Belt bag, Force®8
Catalog # JSPABPT170-011-000
Supplier:  JSP
undefined
Belt bag, Force®8
Catalog # JSPABPT170-011-000
Supplier:  JSP
Specifications
  • Description:
    Force®8 Belt bag, for masks and filters
  • Pk:
    10
Specifications
About this item
Force®8 Belt Bag holds mask and filters when not in use, keeping them free from dirt and damage.
  • Convenient accessibility for Force®8 half-masks
  • Filters and mask remain clean and safe when in bag
  • Increased lifespan for stored items
This robust bag is secured with a wide, comfortable strap and string clip. The main compartment allows for the storage of the Force®8 half-mask without the filters attached. On either side of this main compartment are two more compartments suitable for a filter each. This keeps a complete unit safe and clean.