Impact goggle, Martcare®
Catalog # JSPAAGC010-301-300
Supplier: JSP
Specifications
- Description:Martcare® impact goggle
- Lenses:Polycarbonate
- Pk:1
Specifications
About this item
A cost-effective, compact, and lightweight goggle.
- Compact and lightweight
- Tough polycarbonate lens
- Impact protection
- Vented body
- PVC frame
A compact and lightweight goggle with tough polycarbonate lens and PVC frame, offering high resistance to impact from flying particles.
Certifications: Conforms to EN 166.1.B.