PU banded ear plugs, Soundstopper™
Catalog # JSPAAEE090-070-21X
Supplier: JSP
Specifications
- Description:PU foam plugs with PP headband, SNR 20
- Pk:1
Specifications
About this item
Lightweight and comfortable PU foam earplugs are designed to be worn banded under the chin, ideal for situations where they need to be accessible for long periods of time.
- Lightweight and comfortable to use
- Mould easily to achieve comfortable fit
- SNR: 20
- Band is adjustable for added comfort
These PU foam ear plugs can be conveniently stored around the neck when not in use. Lightweight and comfortable to use over prolonged periods. Easy to fit non-invasive design (earplugs do not penetrate the ear canal).
Certifications: Conforms to EN 352-2.