OverSpecs, swiss one, Coverlite™
Catalog # JSPA1COV23C
Supplier: JSP
Specifications
- Description:Coverlite™ swiss one overspecs
- Seq:01
- Lenses:Clear
- Pk:1
Specifications
About this item
Overspecs with an unrestricted peripheral vision, ideal for prescription wearers or for use as a visitor's spectacle.
- Unrestricted peripheral vision
- Lateral protection
- Soft temple tips
- Ventilated browguard
- Pressure-free fit
Coverlite™ offers one of the best definitions of creative eyewear, overspecs for prescription wearers or spectacle for visitors. With an unrestricted peripheral vision, high side arms graduate to soft, flexible material for additional comfort, pantoscopic side arms adapt to any facial shape for customised fit, and optimal protection with soft grip for a pressure-free fit.
Certifications: Meets EN 166:2001.