Order Entry
Germany
ContactUsLinkComponent
VWR®, Phosphate Buffered Saline (PBS), pH 7,4
VWR®, Phosphate Buffered Saline (PBS), pH 7,4
Catalog # 392-0442
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Phosphate Buffered Saline (PBS), pH 7,4
Catalog # 392-0442
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    PBS without calcium and magnesium X1
  • pH value:
    7,4

Specifications

About this item

Phosphate Buffered Saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate and potassium chloride. PBS helps to maintain a constant pH, is isotonic and non toxic to cells, and provide cells with water and essential inorganic ions. This formulation does not contain calcium and magnesium.

  • PBS is used in a variety of laboratory applications:
  • Washing cells - prior to DNA/RNA extraction
  • Reagent preparation/dilution
  • Transporting cells
  • Diluent of human biological samples

Note: Please refer to the technical data sheet for recommended use.