ContactUsLinkComponent
Digital saccharimeters, ADS400 series
Digital saccharimeters, ADS400 series
Supplier:  BELLINGHAM STANLEY
Certificates
undefined
Digital saccharimeters, ADS400 series
Supplier:  BELLINGHAM STANLEY
Specifications
  • Catalog No:
  • BELL37-25
  • BELL37-26
  • BELL37-85
  • BELL37-27
  • BELL37-86
  • BELL37-87
  • Model:
  • ADS435
  • ADS435-Flow100
  • ADS438
  • ADS435-Flow200
  • ADS438-Flow100
  • ADS438-Flow200
  • Sugar scale accuracy:
  • ±0,030
  • ±0,030
  • ±0,060
  • ±0,030
  • ±0,060
  • ±0,060
  • Sugar scale range:
  • −225...+225 °Z
  • −225...+225 °Z
  • −225...+225 °Z
  • −225...+225 °Z
  • −225...+225 °Z
  • −225...+225 °Z
  • Resolution:
  • 0,001
  • 0,001
  • 0,001
  • 0,001
  • 0,001
  • 0,001
  • Reproducibility:
  • ±0,005
  • ±0,005
  • ±0,010
  • ±0,005
  • ±0,010
  • ±0,010
  • Light source:
  • LED (100,000+ hours)
  • LED (100,000+ hours)
  • LED (100,000+ hours)
  • LED (100,000+ hours)
  • LED (100,000+ hours)
  • LED (100,000+ hours)
  • Interfaces:
  • 1× USB (A), 1× USB (B), 1× Ethernet
  • 1× USB (A), 1× USB (B), 1× Ethernet
  • 1× USB (A), 1× USB (B), 1× Ethernet
  • 1× USB (A), 1× USB (B), 1× Ethernet
  • 1× USB (A), 1× USB (B), 1× Ethernet
  • 1× USB (A), 1× USB (B), 1× Ethernet
  • Temperature range:
  • 5...40 °C
  • 5...40 °C
  • 5...40 °C
  • 5...40 °C
  • 5...40 °C
  • 5...40 °C
  • Optical density range (ext.Ø):
  • 0,0…3,0
  • 0,0…3,0
  • 0,0…3,0
  • 0,0…3,0
  • 0,0…3,0
  • 0,0…3,0
  • User interface:
  • 4" colour with tactile keys
  • 4" colour with tactile keys
  • 4" colour with tactile keys
  • 4" colour with tactile keys
  • 4" colour with tactile keys
  • 4" colour with tactile keys
Specifications