Orders LinkContactUsLinkComponent
Multicollector, b.safe
Multicollector, b.safe
Supplier:   Bohlender
Multicollector, b.safe
Supplier:  Bohlender
Certificates
Multicollector, b.safe
Multicollector, b.safe
2 Options Available Below
Product Details & Documents
Combination of capillaries and tubes of several HPLC systems in one desktop mount.
  • With a b.safe Multicollector you simply multiply the number of connectors of a b.safe Desktop Mount
  • This extension can be done without any modification of the existing installation
  • Chemically inert
  • Static dissipative to prevent static charging
Made of static dissipative PE-EX, screw cap made of PPS with GL 45 thread, rotary insert of static dissipative PE-EX and male threads GL 45. To connect up to 4 b.safe System Caps GL 45 to one desktop mount GL 45 (not included in scope of delivery).
Product Family Options
Product Information
  • Thread
    Availability
    Price
  • BOHLR1140-03
    Multicollector
    Multicollector
    3× GL 45 (male), GL 45 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €
  • BOHLR1140-04
    Multicollector
    Multicollector
    4× GL 45 (male), GL 45 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 173,00 €
    173,00 €
    1 * 1 ST
       173,00 €
Bore adapter
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Bore adapter 75×30 mm, bore in tabletop max. 54 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 73,80 €
    73,80 €
    1 * 1 ST
       73,80 €