Blind cap for waste pipes, b.safe
Catalog # BOHLR1160-05
Supplier: Bohlender
Specifications
- Description:Blind cap for waste pipes, PE, for thread M38×2
- Pk:1
Specifications
About this item
Cap made of PE-EX with pressed-in conical sealing plate made of static-dissipative PTFE-EX.
Suitable sealing cap for clamp connections for tube Ext.Ø 32 mm.