ContactUsLinkComponent
Waste Tubing Desktop Mounts, b.safe Multiport IV
Waste Tubing Desktop Mounts, b.safe Multiport IV
Catalog # BOHLR1127-04
Supplier:  Bohlender
undefined
Waste Tubing Desktop Mounts, b.safe Multiport IV
Catalog # BOHLR1127-04
Supplier:  Bohlender
Specifications
  • Length:
    105 mm
  • Pk:
    1
Specifications
About this item
One connection for up to four HPLC systems that can be integrated directly in laboratory hoods or working tables. b.safe Waste Tubes lead liquid solvent waste directly into connected canisters.
  • liquid solvent wastes can be led directly from the HPLC device into the collecting canister
  • The single pipe is connected by simply pushing it into the lock nut of the joining piece
  • The combination possibilities for the connectors, like e.g. elbow, pipe bend, allow many different installations
Made of static dissipative PE-EX, suitable for tiles with a thickness of 10 to 35 mm, easy fixing by counter nut. Low installation height due to connections at the sides, all threads are inclined to provide additional sloping and connection of tubes without bending. Connections at upper side: 4×2 capillaries (female thread UNF ¹/₄") to connect capillaries with b.safe fittings, 4×1 tube connection (female thread NPT ¹/₈") for hose connectors. Connections at bottom: Ext.Ø 32 mm for direct connection with rigid b.safe Pipe Connections.
Ordering information: Fittings and hose connectors are not included in the scope of delivery.