Modular vacuum concentrators, Savant™ SpeedVac™
Supplier: THERMO SAVANT
Certificates
About this item
The Thermo Scientific™ Savant™ SpeedVac™ SPD210 vacuum concentrator is a large-capacity vacuum concentrator with a modular design, ideal for drying aqueous and organic solvents.
- Kit
- Medium capacity
- Variety of rotors
- Glass safety cover
- PTFE coated aluminum chamber
The SpeedVac™ SPD210 is capable of drying larger quantities of samples at one time and will support your high-volume sample preparation needs.
Ordering information: Supplied as different packages, please see description for details of what is included in specific kits/packages.