Orders LinkContactUsLinkComponent
Cryogenic Lamination Tapes
Cryogenic Lamination Tapes
Supplier:   GA
Cryogenic Lamination Tapes
Supplier:  GA
Certificates
Cryogenic Lamination Tapes
Cryogenic Lamination Tapes
3 Options Available Below
Product Details & Documents
Thin cryo lamination tape provides added protection against abrasion, harsh chemicals, and extreme temperatures. Designed to withstand long-term storage in liquid nitrogen and ultra-low temperature freezers, multiple freeze-thaw cycles, and transport on dry ice. This clear tape can be applied over existing labels.
  • Temperature range: −196 to +93 °C
  • Clear lamination tape protects material from extreme cryogenic conditions, including liquid phase and vapour phase liquid nitrogen
  • Resists harsh laboratory environments, including multiple freeze-thaw cycles
  • Offers protection against chemicals and solvents
  • Resistant to gamma irradiation (tested up to 50 kGy)
This ultra-thin cryo laminating tape provides excellent protection against most chemicals and reagents. It has also been designed to withstand long-term storage in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers (as low as –196 °C) when applied on cryo vials and tubes. Resistant to dry ice, boiling water, multiple freeze-thaw cycles, and other harsh laboratory environments.
Product Family Options
Product Information
  • Colour
    Length
    Width
    Availability
    Price
  • GAINCLAM-19C3-200
    Cryogenic lamination tape, core size: 76,2 mm
    Cryogenic lamination tape, core size: 76,2 mm
    Transparent
    60 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,30 €
    35,30 €
    1 * 1 Rolle
       35,30 €
  • GAINCLAM-25C3-200
    Cryogenic lamination tape, core size: 76,2 mm
    Cryogenic lamination tape, core size: 76,2 mm
    Transparent
    60 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 45,30 €
    45,30 €
    1 * 1 Rolle
       45,30 €
  • GAINCLAM-50C3-200
    Cryogenic lamination tape, core size: 76,2 mm
    Cryogenic lamination tape, core size: 76,2 mm
    Transparent
    60 m
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 55,80 €
    55,80 €
    1 * 1 Rolle
       55,80 €