Graphene oxide 4-10% edge-oxidized, powder 15-20 sheets, Sigma-Aldrich®
Catalog # SIAL796034-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Density:~1.8 g/cm3
:M-Clarity™ quality level = MQ100
Formula:CxOyHz