Order Entry
Germany
ContactUsLinkComponent
1,1,3,3-Tetramethyldisiloxane 97%
1,1,3,3-Tetramethyldisiloxane 97%
Catalog # ABCRAB133126.0100
Supplier:  abcr
CAS Number:  
1,1,3,3-Tetramethyldisiloxane 97%
Catalog # ABCRAB133126.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :134.33 g/mol
 :GHS02
CAS Number:3277-26-7
Formula:C₄H₁₄OSi₂
MDL Number:MFCD00008256