ContactUsLinkComponent
Cellulose chromatography and blotting paper roll, Whatman™
Cellulose chromatography and blotting paper roll, Whatman™
Catalog # WHAT10539028
undefined
Cellulose chromatography and blotting paper roll, Whatman™
Catalog # WHAT10539028
Specifications
  • Size:
    3.8 cm × 137.1 m
  • Pk:
    1
Specifications
About this item
Cellulose Chromatography and Blotting Paper Roll contains cotton linter filter sheets for electrophoresis and amino acid chromatography.
It is also ideal for use with gelatinous samples.