About this item
Save time and standardise activities in equipment preparation for sterilisation. Leave in place during stopper bowl installation to prevent microbial and particulate contamination, and while remaining fill line set-up activities are performed.
- Manufactured and packed in a controlled cleanroom environment
- Reduces labour costs
- Individually poly bagged
- Simplifies conformance when wrapping to SOP's
- Steam sterilisation indicator - autoclavable at 121 to 125 °C
- Elastic to fit a range of diameters
- Tear and puncture resistant, hydrophobic, and lint-free Tyvek® 1422A
Stopper bowl covers provide a convenient, time-saving method of wrapping stopper bowls prior to steam sterilisation. These covers allow immediate steam penetration and work best with modern pulse vacuum autoclaves for sufficient drying.
After sterilisation, and after the outer Tyvek® bag is removed, leave cover in place during installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed. Remaining in place over the bowl during set-up, their use minimises microbial or particulate contamination risks.