Orders LinkContactUsLinkComponent
Swing samplers
Swing samplers
Supplier:   NASCO FORT ATKINSON
Swing samplers
Certificates
Swing samplers
2 Options Available Below
Product Details & Documents
For collecting liquids, powders, or small solids from a horizontal flowing stream
The end of the sampler swings to allow collection from different angles, including 90°. Samplers come with an extending fiberglass pole and one 960mL (32.5oz.) polyethylene bottle with plastic snapper ring.
Product Family Options
Product Information
  • Availability
    Price
  • NASCB01310WA
    Swing sampler, up to 365 cm
    Swing sampler, up to 365 cm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 623,00 €
    623,00 €
    1 * 1 ST
       623,00 €
  • NASCB01366WA
    Swing sampler, up to 730 cm
    Swing sampler, up to 730 cm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 799,00 €
    799,00 €
    1 * 1 ST
       799,00 €