Microvolume UV/visible spectrophotometer, NanoDrop™ Eight
Catalog # 732-3808
Supplier: THERMO NANODROP
Specifications
- Sample size:1 µl
- Description:Microvolume UV/VIS spectrophotometer
- Pk:1
Specifications
About this item
The NanoDrop Eight 8-channel UV/VIS spectrophotometer is the next generation of the NanoDrop 8000 with improved hardware and the best software features of the Thermo Scientific™ NanoDrop™ One/OneC (Acclaro Contaminant Identification) spectrophotometer.
- Measure eight samples in just 20 seconds: 96 samples in less than 6 minutes
- Integrated autostart function for the simplest operation and highest measurement speed
- Acclaro™ module for fast and precise absorbance spectrum analysis and identification of sample impurities
- Real-time identification and correction of DNA/RNA contamination
- Patented sample holding system enables absorbance measurements of up to 10,000 ng/µl dsDNA in a sample volume of only 1 µl without cuvettes
Preconfigured methods for DNA, Protein A280, Microarray, Proteins and Labeling, Pierce 660, Bradford, BCA and Lowry.