Anti-Nuclear Pore Complex Proteins Hashtag 11 Mouse Monoclonal Antibody [clone: MAb414] (TotalSeq™-A0461)
Catalog # BLEG682225
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:Nuclear Pore Complex Proteins Hashtag 11
- Clonality:Monoclonal
- Clone:MAb414
- Conjugation:TotalSeq™-A0461
- Host:Mouse
- Isotype:IgG1
- Reactivity:Yeast,Xenopus,Vertebrates
- Antigen synonyms:Nuclear pore complex protein Nup107|NUP98|Nucleoporin Nup107|NUP84|NPC proteins|NUP107|NUP 107|Nucleoporin 107kD|NUP153|NUP155|107 kD nucleoporin|NUP 84
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:The antibody was raised using a nuclear pore complex mixture.
- Tested applications:PG
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-A oligomer under optimal conditions.
- Size:10 µg
- Pk:10 µG