Anti-CD119 (IFN-gamma R alpha chain) Mouse Monoclonal Antibody [clone: GIR-208] (TotalSeq™-B0219)
Catalog # BLEG308611
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:CD119 (IFN-gamma R alpha chain)
- Clonality:Monoclonal
- Clone:GIR-208
- Conjugation:TotalSeq™-B0219
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,African green monkey,Cynomolgus monkey,Rhesus monkey,Human
- Antigen synonyms:IFN-γR|IFN-α R1|IFN-γRα|CDw119
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:Human IFN-γRα, Purified from human placenta
- Tested applications:PG
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-B oligomer under optimal conditions.
- Size:10 µg
- Pk:10 µG