Anti-CD90.1 (Thy-1.1) Mouse Monoclonal Antibody [clone: OX-7] (TotalSeq™-B0380)
Catalog # BLEG202549
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:CD90.1 (Thy-1.1)
- Clonality:Monoclonal
- Clone:OX-7
- Conjugation:TotalSeq™-B0380
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Rabbit,Guinea pig,Rat,Mouse
- Antigen synonyms:Thy-1|Thy-1.1
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:Rat thymocyte Thy-1 antigen
- Tested applications:PG
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-B oligomer under optimal conditions.
- Size:10 µg
- Pk:10 µG