Anti-CD81 (TAPA-1) Mouse Monoclonal Antibody [clone: 5A6] (TotalSeq™-B0373)
Catalog # BLEG349525
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:CD81 (TAPA-1)
- Clonality:Monoclonal
- Clone:5A6
- Conjugation:TotalSeq™-B0373
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,African green monkey,Cynomolgus monkey,Rhesus monkey,Human
- Antigen synonyms:TAPA-1 (target of an antiproliferative antibody)
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:Human OCI-LY8 cell line
- Tested applications:PG
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-B oligomer under optimal conditions.
- Size:10 µg
- Pk:10 µG