Anti-CD112 (Nectin-2) Mouse Monoclonal Antibody [clone: TX31] (TotalSeq™-C0024)
BLEG337419
- Antigen symbol:CD112 (Nectin-2)
- Clonality:Monoclonal
- Clone:TX31
- Conjugation:TotalSeq™-C0024
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen synonyms:Poliovirus Receptor Related 2 Protein (PRR2)|Nectin-2|Hve B
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA.
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Tested applications:PG
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-C oligomer under optimal conditions.
- Size:10 µg
- Pk:10 µG