Anti-Synapsin I/II/III Mouse Monoclonal Antibody [clone: A17080A] (Biotin)
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG853706
- BLEG853705
- Antigen symbol:
- Synapsin I/II/III
- Synapsin I/II/III
- Antigen synonyms:
- Synapsin III|Synapsin II|Synapsin I|Synapsin|SYN
- Synapsin III|Synapsin II|Synapsin I|Synapsin|SYN
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- A17080A
- A17080A
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant protein corresponding to amino acids 362-450 of human Synapsin I
- Recombinant protein corresponding to amino acids 362-450 of human Synapsin I
- Purification:
- The antibody was purified by affinity chromatography and conjugated with biotin under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with biotin under optimal conditions.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- WB, IHC-P
- WB, IHC-P