Anti-HLA-DR, DP, DQ Mouse Monoclonal Antibody [clone: Tü39] (APC (Allophycocyanin)/FIRE™750)
- Catalog No:
- BLEG361712
- BLEG361711
- Antigen symbol:
- HLA-DR, DP, DQ
- HLA-DR, DP, DQ
- Antigen synonyms:
- human leukocyte antigen|Major Histocompatibility complex II|MHC class II|HLA
- human leukocyte antigen|Major Histocompatibility complex II|MHC class II|HLA
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)/FIRE™750
- APC (Allophycocyanin)/FIRE™750
- Clone:
- Tü39
- Tü39
- Reactivity:
- Baboon, Cynomolgus monkey, Human, Bovine, Dog, Cat
- Baboon, Cynomolgus monkey, Human, Bovine, Dog, Cat
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human PBL
- Human PBL
- Purification:
- The antibody was purified by affinity chromatography and conjugated with APC/Fire™ 750 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with APC/Fire™ 750 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC