Anti-CD274 (B7-H1, PD-L1) Mouse Monoclonal Antibody [clone: MIH2] (APC (Allophycocyanin))
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG393609
- BLEG393610
- Antigen symbol:
- CD274 (B7-H1, PD-L1)
- CD274 (B7-H1, PD-L1)
- Antigen synonyms:
- Programmed cell death ligand 1|PDL1|B7-H1|PD-L1|B7H1|B7 homolog 1|B7-H|PDCD1L1|PDCD1LG1
- Programmed cell death ligand 1|PDL1|B7-H1|PD-L1|B7H1|B7 homolog 1|B7-H|PDCD1L1|PDCD1LG1
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- MIH2
- MIH2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human PD-L1-transfected cells
- Human PD-L1-transfected cells
- Purification:
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC