Order Entry
Germany
ContactUsLinkComponent
Anti-CD105 (Endoglin) Mouse Monoclonal Antibody [clone: SN6h] (Brilliant Violet® 421)
Anti-CD105 (Endoglin) Mouse Monoclonal Antibody [clone: SN6h] (Brilliant Violet® 421)
Supplier:  BIOLEGEND INC
Certificates
Anti-CD105 (Endoglin) Mouse Monoclonal Antibody [clone: SN6h] (Brilliant Violet® 421)
Supplier:  BIOLEGEND INC

Specifications

  • Catalog No:
  • BLEG800509
  • BLEG800510
  • Antigen symbol:
  • CD105 (Endoglin)
  • CD105 (Endoglin)
  • Antigen synonyms:
  • Edg|ENG|END|ORW1|Cell surface MJ7/18 antigen|HHT1
  • Edg|ENG|END|ORW1|Cell surface MJ7/18 antigen|HHT1
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Brilliant Violet® 421
  • Brilliant Violet® 421
  • Clone:
  • SN6h
  • SN6h
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • CD105 (Endoglin) isolated from acute lymphoblastic leukemia cells.
  • CD105 (Endoglin) isolated from acute lymphoblastic leukemia cells.
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • FC
  • FC

Specifications