Anti-CD144 (VE-Cadherin) Mouse Monoclonal Antibody [clone: BV9] (PE (Phycoerythrin)/Cy7®)
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG348515
- BLEG348516
- Antigen symbol:
- CD144 (VE-Cadherin)
- CD144 (VE-Cadherin)
- Antigen synonyms:
- CDH5|VE-Cadherin|cadherin-5
- CDH5|VE-Cadherin|cadherin-5
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Cy7®
- PE (Phycoerythrin)/Cy7®
- Clone:
- BV9
- BV9
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Cyanine7 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE/Cyanine7 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC