Anti-MYDGF (SF20) Rat Monoclonal Antibody [clone: W16077A ]
Catalog # BLEG695802
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:MYDGF (SF20)
- Clonality:Monoclonal
- Clone:W16077A
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Format:Purified
- Antigen synonyms:Myeloid-derived growth factor|Interleukin-25 (IL-25)|Stromal cell-derived growth factor SF20,
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:Partial human MYDGF recombinant protein (32-173 a.a.) expressed in E. coli.
- Tested applications:WB, ICC, KO/KD-WB
- Purification:The antibody was purified by affinity chromatography.
- Size:100 µg
- Pk:100 µG