Anti-CD11c Mouse Monoclonal Antibody [clone: S-HCL-3] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG371518
- BLEG371517
- Antigen symbol:
- CD11c
- CD11c
- Antigen synonyms:
- Integrin αX subunit|ITGAX|CR4|p150
- Integrin αX subunit|ITGAX|CR4|p150
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Clone:
- S-HCL-3
- S-HCL-3
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Spleen cells from patient diagnosed with hairy cell leukemia.
- Spleen cells from patient diagnosed with hairy cell leukemia.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cyanine5.5 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cyanine5.5 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC