Anti-GPR15 Mouse Monoclonal Antibody [clone: SA302A10] (PE (Phycoerythrin))
- Catalog No:
- BLEG373004
- BLEG373003
- Antigen symbol:
- GPR15
- GPR15
- Antigen synonyms:
- BOB|Brother of Bonzo
- BOB|Brother of Bonzo
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- SA302A10
- SA302A10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human GPR15-transfected cells.
- Human GPR15-transfected cells.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC