Anti-NOS2 Mouse Monoclonal Antibody [clone: 5C1B52]
BLEG690902
- Antigen symbol:NOS2
- Clonality:Monoclonal
- Clone:5C1B52
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Format:Purified
- Antigen synonyms:inducible nitric oxide synthase (iNOS)|HEP-NOS|NOS2A|Nitric oxide synthase 2|nitric oxide synthase 2A
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:Partial human NOS2 recombinant protein (1-200 a.a.) expressed in E. coli.
- Tested applications:WB, ICC
- Purification:The antibody was purified by affinity chromatography.
- Size:100 µg
- Pk:100 µG