Anti-Tudor-SN Mouse Monoclonal Antibody [clone: 13.14.1]
BLEG690002
- Antigen symbol:Tudor-SN
- Clonality:Monoclonal
- Clone:13.14.1
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Format:Purified
- Antigen synonyms:Staphylococcal nuclease domain-containing protein 1|p100|SND1|TDRD11|100 kD coactivator
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:C-term region (698-910 a.a.)
- Tested applications:WB, KO/KD-WB
- Purification:The antibody was purified by affinity chromatography.
- Size:100 µg
- Pk:100 µG