Anti-Loricrin Rabbit Polyclonal Antibody [clone: Poly19051]
- Catalog No:
- BLEG905103
- BLEG905104
- Antigen symbol:
- Loricrin
- Loricrin
- Antigen synonyms:
- Loricrin
- Loricrin
- Clonality:
- Polyclonal
- Polyclonal
- Clone:
- Poly19051
- Poly19051
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- This antibody was raised against a peptide sequence derived from the C-terminus of the mouse loricrin protein.
- This antibody was raised against a peptide sequence derived from the C-terminus of the mouse loricrin protein.
- Format:
- Purified
- Purified
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- IHC-P, WB
- IHC-P, WB