Anti-S100A4 Mouse Monoclonal Antibody [clone: NJ-4F3-D1] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
- Catalog No:
- BLEG370009
- BLEG370010
- Antigen symbol:
- S100A4
- S100A4
- Antigen synonyms:
- P9KA|CAPL|Placental calcium-binding protein|Calvasculin|PEL98|FSP1|S100 Calcium Binding Protein A4|Metastasin|MTS1|18A2|42A
- P9KA|CAPL|Placental calcium-binding protein|Calvasculin|PEL98|FSP1|S100 Calcium Binding Protein A4|Metastasin|MTS1|18A2|42A
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Clone:
- NJ-4F3-D1
- NJ-4F3-D1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- cDNA
- cDNA
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cyanine5.5 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cyanine5.5 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC
- ICFC