Anti-A2B5 Mouse Monoclonal Antibody [clone: 105] (Alexa Fluor® 647)
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG150704
- BLEG150703
- Antigen symbol:
- A2B5
- A2B5
- Antigen synonyms:
- F12A2B5|105
- F12A2B5|105
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 647
- Alexa Fluor® 647
- Clone:
- 105
- 105
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgM kappa
- IgM kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Embryonic chicken retinal cells.
- Embryonic chicken retinal cells.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together