Order Entry
Germany
ContactUsLinkComponent
Anti-Arginase 1 Mouse Monoclonal Antibody [clone: O94E6/ARG1]
Anti-Arginase 1 Mouse Monoclonal Antibody [clone: O94E6/ARG1]
Supplier:  BIOLEGEND INC
Certificates
Anti-Arginase 1 Mouse Monoclonal Antibody [clone: O94E6/ARG1]
Supplier:  BIOLEGEND INC

Specifications

  • Catalog No:
  • BLEG678801
  • BLEG678802
  • Antigen symbol:
  • Arginase 1
  • Arginase 1
  • Antigen synonyms:
  • Type I Arginase|liver type Arginase|Arginase 1|ARG1
  • Type I Arginase|liver type Arginase|Arginase 1|ARG1
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • O94E6/ARG1
  • O94E6/ARG1
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • Immunogen:
  • Full length recombinant human ARG1 protein produced in HEK293T cells.
  • Full length recombinant human ARG1 protein produced in HEK293T cells.
  • Format:
  • Purified
  • Purified
  • Purification:
  • The antibody was purified by affinity chromatography.
  • The antibody was purified by affinity chromatography.
  • Concentration:
  • 0.5 mg/ml
  • 0.5 mg/ml
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • WB, KO/KD-WB
  • WB, KO/KD-WB

Specifications