Anti-Blimp-1 Rat Monoclonal Antibody [clone: 5E7] (PE (Phycoerythrin))
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG150006
- BLEG150005
- Antigen symbol:
- Blimp-1
- Blimp-1
- Antigen synonyms:
- PRDI-BF1|ZNFPR1A1|B lymphocyte-induced maturation protein-1|PRDM-1|PR domain zinc finger protein 1|BLIMP1|PRDM1
- PRDI-BF1|ZNFPR1A1|B lymphocyte-induced maturation protein-1|PRDM-1|PR domain zinc finger protein 1|BLIMP1|PRDM1
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 5E7
- 5E7
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Amino acids 255-395 from mouse Blimp-1 fused with GST.
- Amino acids 255-395 from mouse Blimp-1 fused with GST.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- Concentration:
- 0.2 mg/ml
- 0.2 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC
- ICFC