Anti-GD3 (MEL-1) Mouse Monoclonal Antibody [clone: R24]
Catalog # BLEG917701
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:GD3 (MEL-1)
- Clonality:Monoclonal
- Clone:R24
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG3
- Reactivity:Human
- Format:Purified
- Antigen synonyms:SIAT8|SIAT8-A|ganglioside GD3 synthase|ganglioside-specific alpha-2,8-polysialyltransferase|GD3S|Alpha-N-acetylneuraminide alpha-2,8-sialyltransferase|alpha-2,8-sialyltransferase 8A|sialytransferase St8Sia I|disialoganglioside (GD3) synthase
- Storage buffer:Phosphate-buffered solution with BSA + 0.1% NaN3.
- Storage temperature:2…8 °C
- Immunogen:This antibody was raised against SK-MEL-28 melanoma cell line.
- Tested applications:IHC
- Purification:The antibody was purified by affinity chromatography.
- Size:1 mL
- Pk:1 mL