Anti-Alpha Smooth Muscle Actin Mouse Monoclonal Antibody [clone: 1A4]
BLEG904601
- Antigen symbol:Alpha Smooth Muscle Actin
- Clonality:Monoclonal
- Clone:1A4
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:alpha 2 actin|aortic smooth muscle|Actin|cell growth-inhibiting gene 46 protein|alpha-cardiac actin|alpha-actin-2|growth-inhibiting gene 46
- Storage buffer:Cell Culture Supernatant (dialysed against 0.05 mol/L Tris/HCl, pH 7.2, and containing 15 mmol/L NaN3).
- Storage temperature:2…8 °C
- Immunogen:This antibody was raised against the N-terminal synthetic decapeptide of alpha-smooth muscle actin coupled to keyhole limpet haemocyanin (KLH).
- Tested applications:IHC, ICC, WB
- Purification:Tissue Culture Supernatant
- Size:250 µl
- Pk:250 µl