Anti-Aph-1a, 245-265 (C terminus) Rabbit Polyclonal Antibody [clone: Poly18231]
Catalog # BLEG823101
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:Aph-1a, 245-265 (C terminus)
- Clonality:Polyclonal
- Clone:Poly18231
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:Azide-free
- Antigen synonyms:presenilin-stabilization factor|anterior pharynx defective 1 homolog A|aph-1alpha|CGI-78|Gamma-secretase subunit APH-1A
- Storage buffer:Phosphate-buffered solution, pH 7.2.
- Storage temperature:2…8 °C
- Concentration:1 mg/ml
- Immunogen:The antibody Aph-1a, 245-265 (Poly18231) was raised against the human Aph-1aL/C-terminal region and recognizes the sequence CRRQEDSRVMVYSALRIPPED of human Aph-1aL.
- Tested applications:WB, IF
- Purification:The antibody was purified by affinity chromatography.
- Size:100 µl
- Pk:100 µl