Anti-Neurofilament H (NF-H), non-phosphorylated Mouse Monoclonal Antibody [clone: SMI 32]
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG801702
- BLEG801701
- Antigen symbol:
- Neurofilament H (NF-H), non-phosphorylated
- Neurofilament H (NF-H), non-phosphorylated
- Antigen synonyms:
- NF-H|neurofilament triplet H protein|200 kD neurofilament protein|Neurofilament heavy polypeptide
- NF-H|neurofilament triplet H protein|200 kD neurofilament protein|Neurofilament heavy polypeptide
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- SMI 32
- SMI 32
- Reactivity:
- Mammals, Human, Rat, Mouse
- Mammals, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Format:
- Azide-free
- Azide-free
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Phosphate-buffered solution (no preservatives or carrier proteins).
- Phosphate-buffered solution (no preservatives or carrier proteins).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- IHC-P, WB, Array Tomography, ICC
- IHC-P, WB, Array Tomography, ICC