Anti-ASC (TMS-1) Mouse Monoclonal Antibody [clone: HASC-71] (PE (Phycoerythrin))
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG653903
- BLEG653904
- Antigen symbol:
- ASC (TMS-1)
- ASC (TMS-1)
- Antigen synonyms:
- PYD and CARD domain-containing protein|CARD5|target of methylation-induced silencing (TMS1)|apoptosis-associated speck-like protein containing a CARD|caspase recruitment domain protein 5
- PYD and CARD domain-containing protein|CARD5|target of methylation-induced silencing (TMS1)|apoptosis-associated speck-like protein containing a CARD|caspase recruitment domain protein 5
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- HASC-71
- HASC-71
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Recombinant human ASC (aa 113-195).
- Recombinant human ASC (aa 113-195).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC
- ICFC