Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: DA-7] (Alexa Fluor® 594)
Catalog # BLEG628406
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:Cytokeratin 18
- Clonality:Monoclonal
- Clone:DA-7
- Conjugation:Alexa Fluor® 594
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen synonyms:Keratin 18|Keratin type I cytoskeletal 18
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:Human breast carcinoma cell line PMC-42
- Tested applications:ICC, IHC-P
- Purification:The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions.
- Size:100 µg
- Pk:100 µG