Anti-CD90.1 (Thy-1.1) Mouse Monoclonal Antibody [clone: OX-7] (Brilliant Violet® 605)
BLEG202537
- Antigen symbol:CD90.1 (Thy-1.1)
- Clonality:Monoclonal
- Clone:OX-7
- Conjugation:Brilliant Violet® 605
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Rabbit,Guinea pig,Rat,Mouse
- Antigen synonyms:Thy-1|Thy-1.1
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Concentration:0.2 mg/ml
- Immunogen:Rat thymocyte Thy-1 antigen
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions.
- Size:50 µg
- Pk:50 µG