Anti-TNF-alpha Mouse Monoclonal Antibody [clone: MAb11] (PerCP (Peridinin-Chlorophyll Protein Complex))
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG502923
- BLEG502924
- Antigen symbol:
- TNF-alpha
- TNF-alpha
- Antigen synonyms:
- Differentiation inducing factor (DIF)|Necrosin|TNFSF2|Cachectin|Tumor necrosis factor-α|Macrophage cytotoxic factor (MCF)
- Differentiation inducing factor (DIF)|Necrosin|TNFSF2|Cachectin|Tumor necrosis factor-α|Macrophage cytotoxic factor (MCF)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- Clone:
- MAb11
- MAb11
- Reactivity:
- Baboon, Pig, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Cat
- Baboon, Pig, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Cat
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- E. coli expressed, recombinant human TNF-α
- E. coli expressed, recombinant human TNF-α
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PerCP under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PerCP under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC, FC
- ICFC, FC