Order Entry
Germany
ContactUsLinkComponent
n-Hexane 95%, AR®, Macron Fine Chemicals™
n-Hexane 95%, AR®, Macron Fine Chemicals™
Catalog # 5414-25
CAS Number:  
n-Hexane 95%, AR®, Macron Fine Chemicals™
Catalog # 5414-25
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    ≥95%
  • Water (KF):
    ≤0,02%
  • Titrable Acid (meq/g) :
    ≤0,0005
  • Residue after Evaporation:
    ≤0,001%
  • Sulfur Compounds:
    ≤0,001%

Specification Test Results

About this item

 :
 :86.18 g/mol
UN:1208
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520